Please login to your online account to display your discounted pricing

Methylep-tolyl sulfoxide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  428150050

 View more versions of this product

Catalog No. AC428150050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Glass bottle
Quantity 5g
Chemical Name or Material Methyl p-tolyl sulfoxide
CAS 934-72-5
Assay Percent Range 98%
Molecular Formula C8H10OS
Formula Weight 154.23g/mol
MDL Number MFCD00075175

Specifications

Chemical Name or Material Methyl p-tolyl sulfoxide
Assay Percent Range 98%
Molecular Formula C8H10OS
Formula Weight 154.23g/mol
MDL Number MFCD00075175