missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyleoxalyl chloride, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC166000500

 View more versions of this product

Catalog No. AC166000500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Oxidation reagent
Specifications

Specifications

Methyleoxalyl chloride
Authentic
1.4179 to 1.4199 (20°C, 589nm)
50mL
97%
ClCOCO2CH3
02, 541
ZXUQEPZWVQIOJE-UHFFFAOYSA-N
methyl 2-chloro-2-oxoacetate
79846
Liquid
1.33g/mL
Glass bottle
Undesignated
5781-53-3
C3H3ClO3
MFCD00000705
methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
COC(=O)C(=O)Cl
122.504
122.51
≥96.0%

Chemical Identifiers

5781-53-3
122.504
ZXUQEPZWVQIOJE-UHFFFAOYSA-N
79846
COC(=O)C(=O)Cl
C3H3ClO3
MFCD00000705
methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
methyl 2-chloro-2-oxoacetate
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents