missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl oxalyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166000250

 View more versions of this product

Catalog No. AC166000250



Description

Description

Oxidation reagent
Specifications

Specifications

Methyl oxalyl chloride
46°C
Glass bottle
1.33
Undesignated
5781-53-3
C3H3ClO3
MFCD00000705
acetic acid, chlorooxo-, methyl ester, chloroglyoxylic acid methyl ester, methyl 2-chloro-2-oxo-acetate, methyl chloro oxo acetate, methyl chlorocarbonyl formate, methyl chloroglyoxylate, methyl chlorooxoacetate, methyl oxalyl chloride, methylchlorooxoacetate, methyloxalylchloride
ZXUQEPZWVQIOJE-UHFFFAOYSA-N
methyl 2-chloro-2-oxoacetate
79846
Liquid
1.3300g/mL
Authentic
1.4179 to 1.4199
118°C to 120°C
25mL
96% min. (GC)
ClCOCO2CH3
02, 541
Solubility in water: decomposes
COC(=O)C(=O)Cl
122.51
122.51
97%

Chemical Identifiers

5781-53-3
122.51
ZXUQEPZWVQIOJE-UHFFFAOYSA-N
79846
COC(=O)C(=O)Cl
C3H3ClO3
MFCD00000705
acetic acid, chlorooxo-, methyl ester, chloroglyoxylic acid methyl ester, methyl 2-chloro-2-oxo-acetate, methyl chloro oxo acetate, methyl chlorocarbonyl formate, methyl chloroglyoxylate, methyl chlorooxoacetate, methyl oxalyl chloride, methylchlorooxoacetate, methyloxalylchloride
methyl 2-chloro-2-oxoacetate
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 227-307-4

SDS Documents