Please login to your online account to display your discounted pricing

Methyleo-toluate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414951000

 View more versions of this product

Catalog No. AC414951000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 1.0700g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Color Clear colorless
Quantity 100g
Chemical Name or Material Methyl o-toluate
CAS 89-71-4
Physical Form Clear colorless liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Formula Weight 150.18g/mol
MDL Number MFCD00008428
Synonym Methyl 2-methylbenzoate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl o-toluate
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Formula Weight 150.18g/mol
MDL Number MFCD00008428
Synonym Methyl 2-methylbenzoate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents