Please login to your online account to display your discounted pricing

Methyleo-toluate, 99%, ACROS Organics™

$36.80 - $36.80

Chemical Identifiers

Chemical Name or Material Methyl o-toluate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Formula Weight 150.18
MDL Number MFCD00008428
Synonym Methyl 2-methylbenzoate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC414951000

  SDS
thermo fisher scientific
AC414951000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC414950250

  SDS
thermo fisher scientific
414950250
Glass bottle 25g Each for $36.80
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 150.18
Quantity 100g
Color Clear colorless
Density 1.0700g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3C6H4COOCH3
Chemical Name or Material Methyl o-toluate
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C9H10O2
CAS 89-71-4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00008428
Synonym Methyl 2-methylbenzoate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.