Please login to your online account to display your discounted pricing

Methyleo-toluate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414950250

 View more versions of this product

Catalog No. AC414950250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 150.18
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 1.0700g/mL
Linear Formula CH3C6H4COOCH3
Packaging Glass bottle
Synonym Methyl 2-methylbenzoate
CAS 89-71-4
Chemical Name or Material Methyl o-toluate
Assay Percent Range 99%
Molecular Formula C9H10O2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00008428
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl o-toluate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Formula Weight 150.18
MDL Number MFCD00008428
Synonym Methyl 2-methylbenzoate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents