Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyleneiminoacetonitrile, 98%

$55.89 - $130.47

Chemical Identifiers

Chemical Name or Material Methyleneiminoacetonitrile
CAS 109-82-0
Molecular Formula C3H4N2
Linear Formula CH2=NCH2CN
Formula Weight 68.08
MDL Number MFCD00001889
Beilstein 1848155
Merck Index 14,6057
Synonym N-Methyleneglycinonitrile; Methyleneaminoacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1716214

  SDS
alfa aesar™
A1716214
25g Each for $55.89

AAA1716222

  SDS
alfa aesar™
A1716222
100g Each for $130.47
Description & Specifications

Specifications

Formula Weight 68.08
CAS 109-82-0
Chemical Name or Material Methyleneiminoacetonitrile
Synonym N-Methyleneglycinonitrile; Methyleneaminoacetonitrile
Quantity 25g
RTECS Number MC1925000
TSCA TSCA
Molecular Formula C3H4N2
MDL Number MFCD00001889
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,6057
Melting Point 128° to 129°C
Assay 98%
Linear Formula CH2=NCH2CN
Health Hazard 1 Danger
Beilstein 1848155
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.