Please login to your online account to display your discounted pricing

Alfa Aesar™ Methylenediphosphonic acid, 97%

Chemical Identifiers

Chemical Name or Material Methylenediphosphonic acid
CAS 1984-15-2
Formula Weight 176
MDL Number MFCD00002139
UN Number UN3261
Beilstein 1708494
Merck Index 14,5792
Synonym Bis(dihydroxyphosphoryl)methane; Medronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3027903

  SDS
alfa aesar™
3027903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3027906

  SDS
alfa aesar™
3027906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3027914

  SDS
alfa aesar™
3027914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176
Chemical Name or Material Methylenediphosphonic acid
Physical Form Crystalline
Assay 97%
MDL Number MFCD00002139
CAS 1984-15-2
UN Number UN3261
Beilstein 1708494
Merck Index 14,5792
Synonym Bis(dihydroxyphosphoryl)methane; Medronic acid
Health Hazard 1 Danger
Melting Point 202° to 204°C
Quantity 1g
Odor Odorless
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number SZ9138000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.