missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylenecyclopentane, TCI America™

Manufacturer:  TCI America M09145ML

 View more versions of this product

Catalog No. M09145ML



Specifications

Specifications

Methylenecyclopentane
Colorless
1528-30-9
MFCD00001421
NFJPEKRRHIYYES-UHFFFAOYSA-N
methylidenecyclopentane
73714
Clear Liquid at 20°C
77°C
5mL
C6H10
3295
C=C1CCCC1
82.146
82.15
≥98.0% (GC)

Chemical Identifiers

1528-30-9
82.146
NFJPEKRRHIYYES-UHFFFAOYSA-N
methylidenecyclopentane
C6H10
MFCD00001421
73714
C=C1CCCC1
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents