missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylenecyclohexane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC325510050

 View more versions of this product

Catalog No. AC325510050


 

Specifications Description & Specifications

Specifications

≥97.5%
96.17
Clear colorless
05, 69
Methylenecyclohexane
C7H12
MFCD00001661
Glass bottle
Liquid
5g
0.8g/mL
C6H10(=CH)
1192-37-6
98%
1.4465 to 1.4505 (20°C, 589nm)
Authentic

Chemical Identifiers

98%
C6H10(=CH)
MFCD00001661
≥97.5%
C7H12
96.17
Methylenecyclohexane

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger