missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylenebis(thioglycolic Acid) 96.0+%, TCI America™

$138.25 - $470.72

Chemical Identifiers

CAS 2068-24-8
Molecular Formula C5H8O4S2
Molecular Weight (g/mol) 196.235
MDL Number MFCD00135949
InChI Key CZEVAMMETWVBOD-UHFFFAOYSA-N
Synonym 2,2'-methylenebis sulfanediyl diacetic acid, 2,2'-methylenebis thio diacetic acid, 2-carboxymethyl thio methyl thio acetic acid, acetic acid, 2,2'-methylenebis thio bis, acetic acid, methylenedithio di, carboxymethyl sulfanyl methyl sulfanyl acetic acid, methylenebis-thioglycollic acid, methylenebisthioglycolic acid, methylenedithiodiacetic acid
PubChem CID 74949
IUPAC Name 2-(carboxymethylsulfanylmethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCSCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M02185G View Documents Tci America
M02185G
5g Each for $138.25
 
M021825G View Documents Tci America
M021825G
25g Each for $470.72
 
Specifications

Chemical Identifiers

2068-24-8
196.235
CZEVAMMETWVBOD-UHFFFAOYSA-N
74949
C(C(=O)O)SCSCC(=O)O
C5H8O4S2
MFCD00135949
2,2'-methylenebis sulfanediyl diacetic acid, 2,2'-methylenebis thio diacetic acid, 2-carboxymethyl thio methyl thio acetic acid, acetic acid, 2,2'-methylenebis thio bis, acetic acid, methylenedithio di, carboxymethyl sulfanyl methyl sulfanyl acetic acid, methylenebis-thioglycollic acid, methylenebisthioglycolic acid, methylenedithiodiacetic acid
2-(carboxymethylsulfanylmethylsulfanyl)acetic acid

Specifications

Undesignated
5g
C5H8O4S2
2,2'-methylenebis sulfanediyl diacetic acid, 2,2'-methylenebis thio diacetic acid, 2-carboxymethyl thio methyl thio acetic acid, acetic acid, 2,2'-methylenebis thio bis, acetic acid, methylenedithio di, carboxymethyl sulfanyl methyl sulfanyl acetic acid, methylenebis-thioglycollic acid, methylenebisthioglycolic acid, methylenedithiodiacetic acid
C(C(=O)O)SCSCC(=O)O
196.235
196.24
≥96.0% (T)
129°C
2068-24-8
MFCD00135949
CZEVAMMETWVBOD-UHFFFAOYSA-N
2-(carboxymethylsulfanylmethylsulfanyl)acetic acid
74949
Crystal-Powder at 20°C
Methylenebis(thioglycolic Acid)
Safety and Handling

Safety and Handling

TSCA : Yes