Please login to your online account to display your discounted pricing

Methylene Violet, Certified Bernthsen, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC414300010

 View more versions of this product

Catalog No. AC414300010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Methylene Violet
Name Note Bernthsen
CAS 225115
Absorption Ratio 0.99 to 1.06
EINECS Number 219-733-4
TSCA TSCA
Purity Grade Extra Pure
Physical Form Powder
Molecular Formula C14H12N2OS
Formula Weight 256.32
MDL Number MFCD00005064
Beilstein 27, II, 463
Synonym C.I. 52041
Infrared Spectrum Authentic
Lambda Max 579 to 584nm
Packaging Glass bottle
Color Dark green
Quantity 1g

Specifications

Chemical Name or Material Methylene Violet
Name Note Bernthsen
CAS 225115
Purity Grade Extra Pure
Molecular Formula C14H12N2OS
Formula Weight 256.32
MDL Number MFCD00005064
Synonym C.I. 52041