Please login to your online account to display your discounted pricing

Methylene Violet, Certified Bernthsen, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methylene Violet
Name Note Bernthsen
CAS 225115
Purity Grade Extra Pure
Molecular Formula C14H12N2OS
Formula Weight 256.32
MDL Number MFCD00005064
Synonym C.I. 52041
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC414300010

  SDS
thermo fisher scientific
AC414300010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methylene Violet
Name Note Bernthsen
CAS 225115
Absorption Ratio 0.99 to 1.06
EINECS Number 219-733-4
TSCA TSCA
Purity Grade Extra Pure
Physical Form Powder
Molecular Formula C14H12N2OS
Formula Weight 256.32
MDL Number MFCD00005064
Beilstein 27, II, 463
Synonym C.I. 52041
Infrared Spectrum Authentic
Lambda Max 579 to 584nm
Packaging Glass bottle
Color Dark green
Quantity 1g