Please login to your online account to display your discounted pricing

Methylene Chloride (With Cyclohexene Preservative/HPLC), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  D1384

 View more versions of this product

Catalog No. D1384

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade HPLC Grade
Boiling Point 39°C
Melting Point −97°C
Formula Weight 84.93
Quantity 4L
Purity Grade Notes Mobile phase for HPLC applications
DOT Information DOT Class 6.1, : Poison
Color Colorless
Assay ≥99.9%
Vapor Pressure 20mmHg at 3502°C
Physical Form Liquid
Density 1.33g/cm³
Refractive Index 1.4200 to 1.4230 (at 25°C)
Titratable Acid ≤0.0003meq/g
Absorbance ≤0.014 AU at 254nm
Fluorescence Background Pass Test
Preservative 40 to 100ppm Cyclohexene
Cleanup Symbol Solvent
Water ≤0.02%
ChemAlert Storage Symbol Blue
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Free Halogens Pass Test
Packaging Glass Bottle
Refractive Index at 25 degC 1.4200 to 1.4230
MDL Number MFCD00000881
Density at 25 degC 1.315 to 1.321g/mL
Residue after Evaporation ≤2ppm
Synonym DCM
Chemical Name or Material Methylene Chloride
Name Note With cyclohexene preservative
Assay Percent Range 99.9% min by GC
Molecular Formula CH2Cl2
CAS 75-09-2

Specifications

Chemical Name or Material Methylene Chloride
Name Note With cyclohexene preservative
CAS 75-09-2
Purity Grade HPLC Grade
Purity Grade Notes Mobile phase for HPLC applications
Assay ≥99.9%
Assay Percent Range 99.9% min by GC
Molecular Formula CH2Cl2
Formula Weight 84.93
MDL Number MFCD00000881
Synonym DCM
Accessories

Accessories for Catalog Number: D1384

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

 

SDS & Safety Documents