Please login to your online account to display your discounted pricing

Methylene Chloride (HPLC), Fisher Chemical


HPLC mobile phase for analytical and preparative separations

Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPD143SK1

 View more versions of this product

Catalog No. D143SK-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.9%
Purity Grade HPLC Grade
Boiling Point 39°C
Melting Point −97°C
Formula Weight 84.93
Quantity 1L
Purity Grade Notes Mobile phase for HPLC applications
DOT Information DOT Class 6.1, : Poison
Vapor Pressure 20mmHg at 3502°C
Physical Form Liquid
Color Colorless
Density 1.33g/cm³
Identification Pass Test
Refractive Index 1.4200 to 1.4230 (at 25°C)
Residue after Evaporation 2ppm max.
Assay Percent Range 99.9% min by GC
Absorbance 0.01 AU max.
Synonym DCM
Health Hazard 3 Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Free Halogens Pass Test
Packaging Glass Bottle
Preservative 15 to 200ppm
Water 0.02% max.
MDL Number MFCD00000881
Fluorescence Background Pass Test
Titratable Acid 0.0003mEq/g max.
Cleanup Symbol Solvent
Health Hazard 2 WARNING!
Chemical Name or Material Methylene Chloride
Name Note Stabilized
Molecular Formula CH2Cl2
CAS 75-09-2
ChemAlert Storage Symbol Blue

Specifications

Chemical Name or Material Methylene Chloride
Name Note Stabilized
CAS 75-09-2
Purity Grade HPLC Grade
Purity Grade Notes Mobile phase for HPLC applications
Physical Form Liquid
Assay ≥99.9%
Assay Percent Range 99.9% min by GC
Molecular Formula CH2Cl2
Formula Weight 84.93
MDL Number MFCD00000881
Synonym DCM
Accessories
 

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!