Please login to your online account to display your discounted pricing

Methylene Chloride, Optima™, Fisher Chemical


High purity and versatile solvent for HPLC, GC, GC/MS and pesticide residue analysis

Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  D1511

 View more versions of this product

Catalog No. D151-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.9%
Purity Grade OPTIMA™
Boiling Point 39°C
Melting Point −97°C
Formula Weight 84.93
Quantity 1L
Purity Grade Notes High purity mobile phase for HPLC and/or extraction solvent for GC applications
DOT Information DOT Class 6.1, : Poison
Vapor Pressure 20mmHg at 3502°C
Certificate of Analysis Certificate of Lot Analysis included with each bottle; UV and other chromatograms available on request.
Physical Form Liquid
Color Colorless
Density 1.33g/cm³
Identification Pass Test
Refractive Index 1.42 to 1.423 (at 25°C)
Water 0.01% max.
MDL Number MFCD00000881
ChemAlert Storage Symbol Blue
Health Hazard 2 WARNING!
Fluorescence Background 1 ppb max.
Packaging Glass Bottle
Preservative 15 to 200ppm
Residue after Evaporation 1ppm max.
Titratable Acid 0.0002mEq/g max.
Absorbance 0.005 AU max. at 254nm
UN Number 1593
Synonym DCM
Pesticide Residue Analysis 10 ng/L max.
Assay Percent Range 99.9% min by GC
Health Hazard 3 Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Free Halogens Pass Test
GC_MS Responsive Substances Pass Test
Chemical Name or Material Methylene Chloride
Cleanup Symbol Solvent
Molecular Formula CH2Cl2
CAS 75-09-2

Specifications

Chemical Name or Material Methylene Chloride
CAS 75-09-2
Purity Grade OPTIMA™
Purity Grade Notes High purity mobile phase for HPLC and/or extraction solvent for GC applications
Assay ≥99.9%
Assay Percent Range 99.9% min by GC
Molecular Formula CH2Cl2
Formula Weight 84.93
MDL Number MFCD00000881
Synonym DCM

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
  • Contains approx. 15- 200 ppm amylene as preservative
  • Also meets ACS specifications
  • Intended for research use only

Accessories
 

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

 

SDS & Safety Documents