Please login to your online account to display your discounted pricing

Methylene Chloride, Low water, BakerDRY™, J.T.Baker™

Specifically manufactured for use in organometallic and oligonucleotide synthesis

$145.78 - $179.20

Chemical Identifiers

Chemical Name or Material Methylene Chloride
CAS 75-09-2
Name Note Low water
Purity Grade BakerDRY™
Molecular Formula CH2Cl2
Formula Weight 84.93
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003493

 
avantor performance
929510
Glass Bottle 100mL Each for $145.78

02003494

 
avantor performance
929512
Narrow Mouth Amber Glass Bottle, w/ Septum 1L Each for $179.20
Description & Specifications

Specifications

Formula Weight 84.93
Chemical Name or Material Methylene Chloride
Specific Gravity 1L = 1.32kg
Molecular Formula CH2Cl2
CAS 75-09-2
Packaging Glass Bottle
Name Note Low water
Purity Grade BakerDRY™
Quantity 100mL