Please login to your online account to display your discounted pricing

Methylene Chloride, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$150.05 - $455.24

Chemical Identifiers

Chemical Name or Material Methylene Chloride
CAS 75-09-2
Purity Grade HPLC Grade
Molecular Formula CH2Cl2
Formula Weight 84.93
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003400

 
avantor performance
931503
Narrow Mouth Amber Glass Bottle 4L Each for $376.78

02003396

 
avantor performance
931502
Narrow Mouth Amber Glass Bottle 1L Each for $150.05

02003401

 
avantor performance
931533
Narrow Mouth PVC Coated Amber Glass Bottle 4L Each for $455.24
Description & Specifications

Specifications

Formula Weight 84.93
Chemical Name or Material Methylene Chloride
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.32kg
Purity Grade HPLC Grade
Quantity 4L
Molecular Formula CH2Cl2
CAS 75-09-2

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use