Please login to your online account to display your discounted pricing

Methylene Chloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$136.06 - $1,860.79

Chemical Identifiers

Chemical Name or Material Methylene Chloride
CAS 75-09-2
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Molecular Formula CH2Cl2
Formula Weight 84.93
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003397

 
avantor performance
932422
Aluminum Bottle 1L Each for $345.78

02003402

 
avantor performance
932403
Narrow Mouth Amber Glass Bottle 4L Each for $653.93

02003403

 
avantor performance
932405
Aluminum Bottle 4L Each for $765.84

02003404

 
avantor performance
932433
Narrow Mouth PVC Coated Amber Bottle 4L Each for $1,860.79

02003406

 
avantor performance
932401
Narrow Mouth Amber Glass Bottle 500mL Each for $136.06
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 84.93
Purity Grade Notes Meets ACS reagent chemical requirements
Physical Form Liquid
Chemical Name or Material Methylene Chloride
CAS 75-09-2
Molecular Formula CH2Cl2
Quantity 1L
Specific Gravity 1L = 1.32kg
Packaging Aluminum Bottle