Please login to your online account to display your discounted pricing

Methylene Blue TS, 0.05% (w/v) Alcoholic, Ricca Chemical

$89.79 - $89.79

Chemical Identifiers

Chemical Name or Material Methylene Blue
Name Note Test Solution
CAS 64-17-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7220-79-3
CAS Min % 0.06
CAS Max % 0.06
CAS 67-56-1 (Methanol (Methyl Alcohol))
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

4862-4

 
ricca chemical company
RI48624
Poly natural 120mL Each for $89.79

4862-16

 
ricca chemical company
RI486216
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear blue
Quantity 120mL
Concentration or Composition Notes 0.05% (w/v)
Solvent or Matrix Alcoholic Solution
Chemical Name or Material Methylene Blue
Name Note Test Solution
CAS 64-17-5
CAS Min % 98.0
Packaging Poly natural
CAS Max % 100.0
CAS 7220-79-3
CAS Min % 0.06
CAS Max % 0.06
CAS 67-56-1 (Methanol (Methyl Alcohol))