missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylene Blue, Reagent Grade, LabChem™

$76.60 - $76.60

Chemical Identifiers

CAS 7732-18-5,7681-11-0,7487-94-7
CAS Max % 93.64,5,1.36
Linear Formula C16H18N3SCl
Formula Weight 319.85
Chemical Name or Material Methylene Blue
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC168508 Lab Chem
LC168508
Amber Glass 25g Each for $76.60
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-11-0,7487-94-7
C16H18N3SCl
Methylene Blue
93.64,5,1.36
319.85
Reagent Grade

Specifications

7732-18-5,7681-11-0,7487-94-7
Powder
319.85
Amber Glass
190°C
25g
Reagent Grade
93.64,5,1.36
C16H18N3SCl
Passes Test
Soluble in water
Greenish
Methylene Blue

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic to aquatic life.

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Call a poison center/doctor if you feel unwell.
Rinse mouth.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

EINECSNumber : 200-515-2

Recommended Storage : Room Temperature