Please login to your online account to display your discounted pricing

Methylene Blue, Loeffler, Certified, LabChem™

$58.14 - $58.14

Chemical Identifiers

Chemical Name or Material Methylene Blue
CAS 7732-18-5
CAS Max % 80.43
CAS 64-17-5
CAS Max % 17.19
CAS 67-56-1
CAS Max % 0.96
CAS 67-63-0
CAS Max % 0.95
CAS 61-73-4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC169601

 
lab chem
LC169601
Poly Bottle 500mL Each for $58.14
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 319.85
Chemical Name or Material Methylene Blue
Quantity 500mL
CAS Max % 0.96
CAS 67-63-0
CAS Max % 0.95
CAS 61-73-4
CAS Max % 0.41
Name Note Loeffler
Physical Form Liquid
Linear Formula C16H18N3SCl
CAS 7732-18-5
Identification Passes Test
Health Hazard 1 Warning
Packaging Poly Bottle
CAS Max % 80.43
Health Hazard 2 GHS H Statement
Suspected of damaging fertility or the unborn child (oral).
CAS 64-17-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
CAS Max % 17.19
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide
CAS 67-56-1
Color Dark blue
Absorbance Passes Test
Traceability to NIST Traceable to NIST
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Suspected of damaging fertility or the unborn child (oral).

Room Temperature