Please login to your online account to display your discounted pricing

Methylene Blue, HARLECO™, EMD Millipore


Quantity 100g

Manufacturer: emd millipore corporation  28416

 View more versions of this product

Catalog No. M28416

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 373.91
Quantity 100g
Color Blue
Purity Grade Certified Biological Stain
Physical Form Liquid
Chemical Name or Material Methylene Blue
Name Note HARLECO™
Molecular Formula C16H18N3S·HCl
Linear Formula C16H18N3S · HCl
CAS 61-73-4
Synonym Swiss Blue

Specifications

Chemical Name or Material Methylene Blue
Name Note HARLECO™
CAS 61-73-4
Purity Grade Certified Biological Stain
Physical Form Liquid
Molecular Formula C16H18N3S·HCl
Linear Formula C16H18N3S · HCl
Formula Weight 373.91
Synonym Swiss Blue