Please login to your online account to display your discounted pricing

Methylene Blue, HARLECO™, EMD Millipore

$339.34 - $339.34

Chemical Identifiers

Chemical Name or Material Methylene Blue
Name Note HARLECO™
CAS 61-73-4
Purity Grade Certified Biological Stain
Physical Form Liquid
Molecular Formula C16H18N3S·HCl
Linear Formula C16H18N3S · HCl
Formula Weight 373.91
Synonym Swiss Blue
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M28416

 
emd millipore corporation
28416
100g Each for $339.34
Description & Specifications

Specifications

Formula Weight 373.91
Quantity 100g
Color Blue
Physical Form Liquid
Purity Grade Certified Biological Stain
Name Note HARLECO™
Chemical Name or Material Methylene Blue
CAS 61-73-4
Molecular Formula C16H18N3S·HCl
Linear Formula C16H18N3S · HCl
Synonym Swiss Blue