missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylene Blue, Fisher BioReagents™

Manufacturer:  Fisher BioReagents FLBP117100

Catalog No. BP117-100




Description

Description

Specifications

Specifications

Methylene Blue
7220-79-3
C16H24ClN3O3S
15, 6132
XQAXGZLFSSPBMK-UHFFFAOYSA-M
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
660 to 664nm
8.0 to 18.0%
Solid
Solubility in water: slight
1.230g/cm³
190°C
3 to 4.5
Vapor Pressure: Negligible
2000g(-1) cm(-1) min.
MFCD00150008
Pass Test
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
373.90
104827
373.9
Glass Bottle
190°C
52015
Green
100g

Chemical Identifiers

7220-79-3
373.90
XQAXGZLFSSPBMK-UHFFFAOYSA-M
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
C16H24ClN3O3S
MFCD00150008
104827
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. Causes eye, skin, and respiratory tract irritation. May cause methemoglobinemia. Ensure adequate ventilation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : RT

SDS Documents
Product Certifications
Promotions

Promotions