missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methylene Blue, 1% Aqueous, Certified, LabChem™
Supplier: LabChem LC169402
Specifications
Passes Test | |
1% Aqueous | |
99,1 | |
C16H18N3SCl | |
Passes Test | |
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-] | |
319.851 | |
CHEBI:6872 | |
Certified | |
1g/mL | |
1g/mL | |
1 L |
Methylene Blue | |
61-73-4,7732-18-5 | |
C16H18ClN3S | |
Soluble in water | |
CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride | |
6099 | |
319.85 | |
Poly Bottle | |
Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds | |
Blue | |
Liquid |
Chemical Identifiers
61-73-4 | |
319.851 | |
6099 | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride |
C16H18ClN3S | |
CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
CHEBI:6872 | |
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature