Please login to your online account to display your discounted pricing

Methylemalonyl chloride, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC414511000

 View more versions of this product

Catalog No. AC414511000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (acid halide)
Physical Form Liquid
Formula Weight 136.53
Quantity 100g
Color Clear light yellow to yellow
Density 1.273g/mL
Beilstein 02, IV, 1886
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C4H5ClO3
Linear Formula ClCOCH2CO2CH3
MDL Number MFCD00013657
Recommended Storage Corrosives
Chemical Name or Material Methylemalonyl chloride
Infrared Spectrum Authentic
Name Note 97%
Refractive Index 1.4310 to 1.4350 (20°C, 589nm)
CAS 37517-81-0
Synonym Methyl 3-chloro-3-oxopropionate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methylemalonyl chloride
Name Note 97%
CAS 37517-81-0
Assay ≥96.0% (acid halide)
Assay Percent Range 97%
Molecular Formula C4H5ClO3
Linear Formula ClCOCH2CO2CH3
Formula Weight 136.53
MDL Number MFCD00013657
Synonym Methyl 3-chloro-3-oxopropionate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Corrosives

SDS & Safety Documents