missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl linoleate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 428130250

 View more versions of this product

Catalog No. AC428130250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl linoleate
-35.0°C
212.0°C (16.0 mmHg)
98.5% min. (GC)
C19H34O2
MFCD00009534
0.89
methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
WTTJVINHCBCLGX-NQLNTKRDSA-N
methyl (9Z,12Z)-octadeca-9,12-dienoate
5284421
294.48
112-63-0
0.8900g/mL
>200°C
Glass bottle
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2CH3
25 g
15, 6167
Solubility in water: insoluble. Other solubilities: miscible with dmf,fat solvents,oils
CCCCCC=CCC=CCCCCCCCC(=O)OC
294.48
CHEBI:69080
99%

Chemical Identifiers

112-63-0
294.48
WTTJVINHCBCLGX-NQLNTKRDSA-N
5284421
methyl (9Z,12Z)-octadeca-9,12-dienoate
C19H34O2
MFCD00009534
methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
CHEBI:69080
CCCCCC=CCC=CCCCCCCCC(=O)OC
Safety and Handling

Safety and Handling

EINECSNumber : 203-993-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only