missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyleisothiocyanate, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C2H3NS
Linear Formula CH3NCS
Formula Weight 73.12
MDL Number MFCD00004818
Synonym MTC
Chemical Name or Material Methyl isothiocyanate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC414471000 SDS Thermo Fisher Scientific
AC414471000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC414470250 SDS Thermo Fisher Scientific
414470250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
CH3NCS
MFCD00004818
Methyl isothiocyanate
C2H3NS
73.12
MTC
≥96.0%

Specifications

Low Melting Solid
100g
15, 6164
30° to 36°C
CH3NCS
Authentic
97%
Glass bottle
≥96.0%
73.12
Pale yellow to yellow-orange
1.0690g/mL
MTC
C2H3NS
556-61-6
MFCD00004818
Methyl isothiocyanate

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger