Please login to your online account to display your discounted pricing

Methyleisothiocyanate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414470250

 View more versions of this product

Catalog No. AC414470250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Low Melting Solid
Formula Weight 73.12
Quantity 25g
Color Pale yellow to yellow-orange
Merck Index 15, 6164
Melting Point 30° to 36°C
Density 1.0690g/mL
Chemical Name or Material Methyl isothiocyanate
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C2H3NS
CAS 556-61-6
Synonym MTC
Packaging Glass bottle
Linear Formula CH3NCS
MDL Number MFCD00004818
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl isothiocyanate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2H3NS
Linear Formula CH3NCS
Formula Weight 73.12
MDL Number MFCD00004818
Synonym MTC
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

SDS & Safety Documents