Please login to your online account to display your discounted pricing

Methyleindole-4-carboxylate, 99%, ACROS Organics™

$66.03 - $66.03

Chemical Identifiers

Chemical Name or Material Methyl indole-4-carboxylate
CAS 39830-66-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00191222
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334030050

  SDS
fisher scientific acros
334030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC334030010

  SDS
thermo fisher scientific
334030010
Glass bottle 1g Each for $66.03
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 175.19
Physical Form Crystalline Powder
Quantity 5g
Color Off-white to green
Melting Point 67° to 71°C
Packaging Glass bottle
Assay Percent Range 99%
CAS 39830-66-5
Chemical Name or Material Methyl indole-4-carboxylate
Molecular Formula C10H9NO2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
MDL Number MFCD00191222
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.