missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl glycolate, 98%, Thermo Scientific Chemicals
$192.01 - $192.01
Chemical Identifiers
CAS | 96-35-5 |
---|---|
Molecular Formula | C3H6O3 |
Molecular Weight (g/mol) | 90.08 |
InChI Key | GSJFXBNYJCXDGI-UHFFFAOYSA-N |
Synonym | methyl glycolate, methyl hydroxyacetate, acetic acid, hydroxy-, methyl ester, glycolic acid, methyl ester, acetic acid, 2-hydroxy-, methyl ester, glycolic acid methyl ester, unii-7g7r729ej0, hydroxyacetic acid methyl ester, glycolic acid methyl, methyl2-hydroxyethanoate |
PubChem CID | 66774 |
IUPAC Name | methyl 2-hydroxyacetate |
SMILES | COC(=O)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC414370250
|
Thermo Scientific Chemicals
414370250 |
25 g | Glass bottle |
Each for $192.01
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-35-5 | |
90.08 | |
methyl glycolate, methyl hydroxyacetate, acetic acid, hydroxy-, methyl ester, glycolic acid, methyl ester, acetic acid, 2-hydroxy-, methyl ester, glycolic acid methyl ester, unii-7g7r729ej0, hydroxyacetic acid methyl ester, glycolic acid methyl, methyl2-hydroxyethanoate | |
methyl 2-hydroxyacetate |
C3H6O3 | |
GSJFXBNYJCXDGI-UHFFFAOYSA-N | |
66774 | |
COC(=O)CO |
Specifications
96-35-5 | |
Colorless | |
52.0°C (17.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
25 g | |
1.184 | |
GSJFXBNYJCXDGI-UHFFFAOYSA-N | |
methyl 2-hydroxyacetate | |
66774 | |
98% | |
Methyl glycolate |
149.0°C to 151.0°C | |
1.1840g/mL | |
67°C | |
97.5% min. (GC) | |
C3H6O3 | |
HOCH2COOCH3 | |
03,236 | |
methyl glycolate, methyl hydroxyacetate, acetic acid, hydroxy-, methyl ester, glycolic acid, methyl ester, acetic acid, 2-hydroxy-, methyl ester, glycolic acid methyl ester, unii-7g7r729ej0, hydroxyacetic acid methyl ester, glycolic acid methyl, methyl2-hydroxyethanoate | |
COC(=O)CO | |
90.08 | |
90.08 | |
Liquid |
Safety and Handling
EINECSNumber : 202-502-7
RUO – Research Use Only