Please login to your online account to display your discounted pricing

Methyleglycolate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC414371000

 View more versions of this product

Catalog No. AC414371000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 90.08
Assay ≥97.5%
Quantity 100g
Color Clear colorless
Density 1.184g/mL
Refractive Index 1.4160 to 1.4180 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material Methyleglycolate
Assay Percent Range 98%
Molecular Formula C3H6O3
CAS 96-35-5
Packaging Glass bottle
Linear Formula HOCH2COOCH3

Specifications

Chemical Name or Material Methyleglycolate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H6O3
Linear Formula HOCH2COOCH3
Formula Weight 90.08