Learn More
Methyl cyclopropanecarboxylate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 126711000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl cyclopropanecarboxylate | |
0.9800g/mL | |
17°C | |
98% | |
C5H8O2 | |
C3H5CO2CH3 | |
100g | |
0.98 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohols | |
COC(=O)C1CC1 | |
100.12 | |
100.12 |
2868-37-3 | |
119.0°C (764.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4170 to 1.4190 | |
MFCD00001280 | |
09, I, 3 | |
methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate | |
PKAHQJNJPDVTDP-UHFFFAOYSA-N | |
methyl cyclopropanecarboxylate | |
76122 | |
98% |
Chemical Identifiers
2868-37-3 | |
100.12 | |
PKAHQJNJPDVTDP-UHFFFAOYSA-N | |
76122 | |
COC(=O)C1CC1 |
C5H8O2 | |
MFCD00001280 | |
methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate | |
methyl cyclopropanecarboxylate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Toxic in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Danger
EINECSNumber : 220-690-9
RUO â Research Use Only