Please login to your online account to display your discounted pricing

Methylecyclobutanecarboxylate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  365540050

 View more versions of this product

Catalog No. AC365540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Formula Weight 114.14
Physical Form Crystalline Powder and Lumps
Quantity 5g
Color White to beige
Infrared Spectrum Authentic
MDL Number MFCD00095142
Molecular Formula C6H10O2
Packaging Glass bottle
Name Note 98%
Refractive Index 1.4260 to 1.4300 (20°C, 589nm)
CAS 765-85-5
Chemical Name or Material Methylecyclobutanecarboxylate
Assay Percent Range 98%

Specifications

Chemical Name or Material Methylecyclobutanecarboxylate
Name Note 98%
CAS 765-85-5
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H10O2
Formula Weight 114.14
MDL Number MFCD00095142