Please login to your online account to display your discounted pricing

Methylecellulose, viscosity 400 cP (2% solution in water), ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428432500

 View more versions of this product

Catalog No. AC428432500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 250g
Name Note Viscosity 400 cP (2% solution in water)
Chemical Name or Material Methyl cellulose
Packaging Glass bottle
MDL Number MFCD00081763
CAS 9004-67-5

Specifications

Chemical Name or Material Methyl cellulose
Name Note Viscosity 400 cP (2% solution in water)
CAS 9004-67-5
MDL Number MFCD00081763