missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle6-methoxy-2-indolecarboxylate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC397920010

 View more versions of this product

Catalog No. AC397920010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

205.21
Powder
White to beige
Consistent with structure
98081-83-5
98%
Glass bottle
≥97.5%
1g
117° to 122°C
C11H11NO3
Methyle6-methoxy-2-indolecarboxylate
MFCD00134301
Authentic

Chemical Identifiers

98081-83-5
C11H11NO3
MFCD00134301
≥97.5%
98%
205.21
Methyle6-methoxy-2-indolecarboxylate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.