Learn More
Methyl 6-methoxy-2-indolecarboxylate, 98%, ACROS Organics™
Chemical Identifiers
CAS | 98081-83-5 |
---|---|
Molecular Formula | C11H11NO3 |
Molecular Weight (g/mol) | 205.21 |
MDL Number | MFCD00134301 |
InChI Key | OPUUCOLVBDQWEY-UHFFFAOYSA-N |
Synonym | 1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester, 6-methoxy-1h-indole-2-carboxylic acid methyl ester, 6-methoxy-2-indolecarboxylate, 6-methoxyindole-2-carboxylic acid methyl ester, methyl 6-methoxy-2-indolecarboxylat, methyl 6-methoxy-2-indolecarboxylate, methyl 6-methoxy-indole-2-carboxylate, methyl 6-methoxyindole-2-carboxylate, methyl-6-methoxy-2-indolecarboxylate, pubchem7251 |
PubChem CID | 688173 |
IUPAC Name | methyl 6-methoxy-1H-indole-2-carboxylate |
SMILES | COC1=CC2=C(C=C1)C=C(N2)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397920010
|
Thermo Fisher Scientific
AC397920010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC397920050
|
Thermo Fisher Scientific
AC397920050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
98081-83-5 | |
205.21 | |
OPUUCOLVBDQWEY-UHFFFAOYSA-N | |
688173 | |
COC1=CC2=C(C=C1)C=C(N2)C(=O)OC |
C11H11NO3 | |
MFCD00134301 | |
1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester, 6-methoxy-1h-indole-2-carboxylic acid methyl ester, 6-methoxy-2-indolecarboxylate, 6-methoxyindole-2-carboxylic acid methyl ester, methyl 6-methoxy-2-indolecarboxylat, methyl 6-methoxy-2-indolecarboxylate, methyl 6-methoxy-indole-2-carboxylate, methyl 6-methoxyindole-2-carboxylate, methyl-6-methoxy-2-indolecarboxylate, pubchem7251 | |
methyl 6-methoxy-1H-indole-2-carboxylate |
Specifications
Authentic | |
Glass bottle | |
117°C to 119°C | |
98081-83-5 | |
C11H11NO3 | |
1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester, 6-methoxy-1h-indole-2-carboxylic acid methyl ester, 6-methoxy-2-indolecarboxylate, 6-methoxyindole-2-carboxylic acid methyl ester, methyl 6-methoxy-2-indolecarboxylat, methyl 6-methoxy-2-indolecarboxylate, methyl 6-methoxy-indole-2-carboxylate, methyl 6-methoxyindole-2-carboxylate, methyl-6-methoxy-2-indolecarboxylate, pubchem7251 | |
COC1=CC2=C(C=C1)C=C(N2)C(=O)OC | |
205.21 | |
205.21 | |
98% |
Consistent with structure | |
White to Beige | |
1g | |
97.5% min. (GC) | |
MFCD00134301 | |
OPUUCOLVBDQWEY-UHFFFAOYSA-N | |
methyl 6-methoxy-1H-indole-2-carboxylate | |
688173 | |
Powder | |
Methyl 6-methoxy-2-indolecarboxylate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning