missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle6-chloropyridazine-4-carboxylate, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC432980010

 View more versions of this product

Catalog No. AC432980010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 6-chloropyridazine-4-carboxylate
Glass bottle
83°C to 90°C
1093860-48-0
C6H5ClN2O2
QYQHGLKSLGUINZ-UHFFFAOYSA-N
methyl 6-chloropyridazine-4-carboxylate
45790080
Liquid or Low Melting Solid
Authentic
Brown to Colorless
1g
97%
6-chloro-pyridazine-4-carboxylic acid methyl ester, methyl6-chloropyridazine-4-carboxylate, 4-pyridazinecarboxylicacid, 6-chloro-, methyl ester, 6-chloro-4-pyridazinecarboxylic acid, methyl ester, 4-pyridazinecarboxylic acid, 6-chloro-, methyl ester
COC(=O)C1=CC(=NN=C1)Cl
172.568
172.57
≥96.0% (GC)

Chemical Identifiers

1093860-48-0
172.568
6-chloro-pyridazine-4-carboxylic acid methyl ester, methyl6-chloropyridazine-4-carboxylate, 4-pyridazinecarboxylicacid, 6-chloro-, methyl ester, 6-chloro-4-pyridazinecarboxylic acid, methyl ester, 4-pyridazinecarboxylic acid, 6-chloro-, methyl ester
methyl 6-chloropyridazine-4-carboxylate
C6H5ClN2O2
QYQHGLKSLGUINZ-UHFFFAOYSA-N
45790080
COC(=O)C1=CC(=NN=C1)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents