Please login to your online account to display your discounted pricing

Methyle6-bromopyridine-2-carboxylate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyle6-bromopyridine-2-carboxylate
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C7H6BrNO2
Formula Weight 216.03
Synonym Methyl 6-bromopicolinate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC436870050

 
thermo fisher scientific
AC436870050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC436870010

 
thermo fisher scientific
AC436870010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (HPLC)
Physical Form Crystalline Powder or Needles
Formula Weight 216.03
Quantity 5g
Color White to pale pink to pale brown to yellow
Melting Point 90° to 96°C
Water 1% Max.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Chemical Name or Material Methyle6-bromopyridine-2-carboxylate
CAS 26218-75-7
Assay Percent Range 97%
Molecular Formula C7H6BrNO2
Synonym Methyl 6-bromopicolinate
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.