missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 6-bromopyridine-2-carboxylate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436870050

 View more versions of this product

Catalog No. AC436870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Methyl 6-bromopyridine-2-carboxylate
Glass bottle
White to Yellow
5g
96% min. (HPLC)
2-bromo-6-methoxycarbonyl pyridine, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 6-bromo-2-pyridine carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, 6-bromopyridine-2-carboxylic acid methyl ester, methyl 2-bromo-6-nicotinate, methyl 6-bromo-2-pyridinecarboxylate, methyl 6-bromopicolinate, methyl-6-bromo-2-pyridine carboxylate, methyl-6-bromopyridine-2-carboxylate
COC(=O)C1=NC(=CC=C1)Br
216.03
216.03
97%
Authentic
1% Max.
94.6°C to 95°C
26218-75-7
C7H6BrNO2
SGNCOKUHMXLGAH-UHFFFAOYSA-N
methyl 6-bromopyridine-2-carboxylate
16217886
Crystalline Powder or Needles

Chemical Identifiers

26218-75-7
216.03
2-bromo-6-methoxycarbonyl pyridine, 2-pyridinecarboxylic acid, 6-bromo-, methyl ester, 6-bromo-2-pyridine carboxylic acid methyl ester, 6-bromopicolinic acid methyl ester, 6-bromopyridine-2-carboxylic acid methyl ester, methyl 2-bromo-6-nicotinate, methyl 6-bromo-2-pyridinecarboxylate, methyl 6-bromopicolinate, methyl-6-bromo-2-pyridine carboxylate, methyl-6-bromopyridine-2-carboxylate
methyl 6-bromopyridine-2-carboxylate
C7H6BrNO2
SGNCOKUHMXLGAH-UHFFFAOYSA-N
16217886
COC(=O)C1=NC(=CC=C1)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

SDS Documents