Please login to your online account to display your discounted pricing

Methyle6-bromo-2-naphthoate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC364140050

 View more versions of this product

Catalog No. AC364140050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 265.1
Quantity 5g
Color Off-white to cream or light brown
Melting Point 118° to 126°C
Assay Percent Range 98%
CAS 33626-98-1
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material Methyl 6-bromo-2-naphthoate
MDL Number MFCD00100408
Name Note 98%
Molecular Formula C12H9BrO2
Linear Formula BrC10H6COOCH3

Specifications

Chemical Name or Material Methyl 6-bromo-2-naphthoate
Name Note 98%
CAS 33626-98-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H9BrO2
Linear Formula BrC10H6COOCH3
Formula Weight 265.1
MDL Number MFCD00100408