Please login to your online account to display your discounted pricing

Methyle6-bromo-2-naphthoate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC364141000

 View more versions of this product

Catalog No. AC364141000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 265.1
Assay ≥97.5%
Quantity 100g
Color Off-white to cream or light brown
Melting Point 118° to 126°C
Linear Formula BrC10H6COOCH3
Molecular Formula C12H9BrO2
MDL Number MFCD00100408
Name Note 98%
Chemical Name or Material Methyl 6-bromo-2-naphthoate
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 33626-98-1
Assay Percent Range 98%

Specifications

Chemical Name or Material Methyl 6-bromo-2-naphthoate
Name Note 98%
CAS 33626-98-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H9BrO2
Linear Formula BrC10H6COOCH3
Formula Weight 265.1
MDL Number MFCD00100408