missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 6-amino-5-bromonicotinate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437940010
Specifications
Methyl 6-amino-5-bromonicotinate | |
Glass bottle | |
173.5°C to 175.0°C | |
180340-70-9 | |
C7H7BrN2O2 | |
3-pyridinecarboxylic acid, 6-amino-5-bromo-, methyl ester, 6-amino-5-bromonicotinic acid methyl ester, acmc-209egj, methyl 2-amino-3-bromo-5-pyridinecarboxylate, methyl 2-amino-3-bromopyridine-5-carboxylate, methyl 6-amino-5-bromo-pyridine-3-carboxylate, methyl 6-amino-5-bromonicotinate, methyl-6-5-bromonicotinate, methyl-6-amino-5-bromonicotinate, pubchem9472 | |
COC(=O)C1=CC(=C(N=C1)N)Br | |
231.05 | |
231.05 | |
98% |
Authentic | |
Brown | |
1g | |
98% | |
MFCD06659561 | |
LNTAAMVZKGYIIA-UHFFFAOYSA-N | |
methyl 6-amino-5-bromopyridine-3-carboxylate | |
2764099 | |
Powder |
Chemical Identifiers
180340-70-9 | |
231.05 | |
LNTAAMVZKGYIIA-UHFFFAOYSA-N | |
2764099 | |
COC(=O)C1=CC(=C(N=C1)N)Br |
C7H7BrN2O2 | |
MFCD06659561 | |
3-pyridinecarboxylic acid, 6-amino-5-bromo-, methyl ester, 6-amino-5-bromonicotinic acid methyl ester, acmc-209egj, methyl 2-amino-3-bromo-5-pyridinecarboxylate, methyl 2-amino-3-bromopyridine-5-carboxylate, methyl 6-amino-5-bromo-pyridine-3-carboxylate, methyl 6-amino-5-bromonicotinate, methyl-6-5-bromonicotinate, methyl-6-amino-5-bromonicotinate, pubchem9472 | |
methyl 6-amino-5-bromopyridine-3-carboxylate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning