Please login to your online account to display your discounted pricing

Methyle6-amino-5-bromonicotinate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyle6-amino-5-bromonicotinate
CAS 180340-70-9
Assay ≥96.0%
Assay Percent Range 98%
Molecular Formula C7H7BrN2O2
Formula Weight 231.05
MDL Number MFCD06659561
Synonym Methyl 6-amino-5-bromopyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC437940010

  SDS
thermo fisher scientific
AC437940010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.05
Assay ≥96.0%
Physical Form Powder
Quantity 1g
Color Brown
Melting Point 172° to 176°C
Synonym Methyl 6-amino-5-bromopyridine-3-carboxylate
Chemical Name or Material Methyle6-amino-5-bromonicotinate
Molecular Formula C7H7BrN2O2
Assay Percent Range 98%
CAS 180340-70-9
MDL Number MFCD06659561
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.