Please login to your online account to display your discounted pricing

Methyle5-formyl-2-methyl-3-furoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 5-formyl-2-methyl-3-furoate
CAS 81661-26-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD03407372
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00666CB

 
fisher scientific company
MO00666CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.15
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C8H8O4
CAS 81661-26-9
Chemical Name or Material Methyl 5-formyl-2-methyl-3-furoate
Packaging Amber Glass Bottle
MDL Number MFCD03407372