Please login to your online account to display your discounted pricing

Methyle5-formyl-2-methyl-3-furoate, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00666CB

 View more versions of this product

Catalog No. MO00666CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 168.15
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C8H8O4
CAS 81661-26-9
Assay Percent Range 97%
MDL Number MFCD03407372
Packaging Amber Glass Bottle
Chemical Name or Material Methyl 5-formyl-2-methyl-3-furoate

Specifications

Chemical Name or Material Methyl 5-formyl-2-methyl-3-furoate
CAS 81661-26-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD03407372