Please login to your online account to display your discounted pricing

Methyle5-fluoro-2-hydroxybenzoate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  398490050

 View more versions of this product

Catalog No. AC398490050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 29° to 34°C
Color Pale yellow
Quantity 5g
Chemical Name or Material Methyl 5-fluoro-2-hydroxybenzoate
CAS 391-92-4
Physical Form Pale yellow low melting solid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H7FO3
Linear Formula FC6H3(OH)COOCH3
Formula Weight 170.14g/mol
MDL Number MFCD03427141
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl 5-fluoro-2-hydroxybenzoate
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H7FO3
Linear Formula FC6H3(OH)COOCH3
Formula Weight 170.14g/mol
MDL Number MFCD03427141
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents