Please login to your online account to display your discounted pricing

Methyle5-fluoro-2-hydroxybenzoate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 5-fluoro-2-hydroxybenzoate
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H7FO3
Linear Formula FC6H3(OH)COOCH3
Formula Weight 170.14g/mol
MDL Number MFCD03427141
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398490050

  SDS
thermo fisher scientific
398490050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 29° to 34°C
Color Pale yellow
Quantity 5g
Chemical Name or Material Methyl 5-fluoro-2-hydroxybenzoate
CAS 391-92-4
Physical Form Pale yellow low melting solid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C8H7FO3
Linear Formula FC6H3(OH)COOCH3
Formula Weight 170.14g/mol
MDL Number MFCD03427141
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.