missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle5-bromo-4-methyl-2-thiophenecarboxylate, 97%, Maybridge

Chemical Identifiers

CAS 54796-47-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C7H7BrO2S
Formula Weight 235.1
MDL Number MFCD04038959
Chemical Name or Material Methyl 5-bromo-4-methyl-2-thiophenecarboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC29010EA SDS Fisher Scientific Company
CC29010EA
Amber Glass Bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC29010DA SDS Fisher Scientific Company
CC29010DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

54796-47-3
100.0
C7H7BrO2S
MFCD04038959
97%
96.5
95%
235.1
Methyl 5-bromo-4-methyl-2-thiophenecarboxylate

Specifications

235.1
100.0
95%
96.5
Amber Glass Bottle
97%
10g
C7H7BrO2S
MFCD04038959
54796-47-3
Methyl 5-bromo-4-methyl-2-thiophenecarboxylate

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.